Tisk…
Anti-CD34 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 4H11(APG)]
Dodavatel: Abcam
ABCAAB18228-50EA 13640 CZK
ABCAAB18228-50
Anti-CD34 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 4H11(APG)]
Protilátky
Type: Primary
Antigen: CD34 (CD34 molecule)
Clonality: monoclonal
Clone: 4H11(APG)
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human