Tisk…

Anti-CD63 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: MEM-259]

Dodavatel: Abcam
ABCAAB18235-50EA 11310 CZK
ABCAAB18235-50
Anti-CD63 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: MEM-259]
Protilátky
Type: Primary
Antigen: CD63 (CD63 molecule)
Clonality: monoclonal
Clone: MEM-259
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR