Tisk…

Anti-NCAM1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-188]

Dodavatel: Abcam
ABCAAB18277-50EA 13120 CZK
ABCAAB18277-50
Anti-NCAM1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-188]
Protilátky
Type: Primary
Antigen: NCAM1 (neural cell adhesion molecule 1)
Clonality: monoclonal
Clone: MEM-188
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR