Tisk…

Anti-CEBPB Mouse monoclonal antibody [clone: A16]

ABCAAB18336-100EA 17450 CZK
ABCAAB18336-100
Anti-CEBPB Mouse monoclonal antibody [clone: A16]
Type: Primary
Antigen: CEBPB (CCAAT/enhancer binding protein (C/EBP), beta)
Clonality: monoclonal
Clone: A16
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR