Tisk…

Anti-FBXO5 Rabbit polyclonal antibody

ABCAAB18341-100EA 15370 CZK
ABCAAB18341-100
Anti-FBXO5 Rabbit polyclonal antibody
Type: Primary
Antigen: FBXO5 (F-box protein 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR