Tisk…

Anti-KRT1 Mouse Monoclonal Antibody [clone: LL017]

Dodavatel: Abcam
ABCAAB18389-50EA 21140 CZK
ABCAAB18389-50
Anti-KRT1 Mouse Monoclonal Antibody [clone: LL017]
Protilátky
Type: Primary
Antigen: KRT1 (keratin 1)
Clonality: monoclonal
Clone: LL017
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR