Tisk…

Anti-IL28 Receptor alpha Goat Polyclonal Antibody

Dodavatel: Abcam
ABCAAB18567-100EA 21850 CZK
ABCAAB18567-100
Anti-IL28 Receptor alpha Goat Polyclonal Antibody
Protilátky
Type: Primary
Antigen: IL28 Receptor alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR