Tisk…

Anti-Rb2 p130 Rabbit polyclonal antibody

ABCAAB18577-100EA 16160 CZK
ABCAAB18577-100
Anti-Rb2 p130 Rabbit polyclonal antibody
Type: Primary
Antigen: Rb2 p130
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR