Tisk…

Anti-C-tag Rabbit polyclonal antibody

ABCAAB18591-200EA 0 CZK
ABCAAB18591-200
Anti-C-tag Rabbit polyclonal antibody
Type: Primary
Antigen: C-tag
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR