Tisk…

Anti-Protein A Chicken Polyclonal Antibody (Biotin)

Dodavatel: Abcam
ABCAAB18598-500EA 22310 CZK
ABCAAB18598-500
Anti-Protein A Chicken Polyclonal Antibody (Biotin)
Protilátky
Type: Primary
Antigen: Protein A
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR