Tisk…

Anti-Protein G Chicken Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Abcam
ABCAAB18599-500EA 22340 CZK
ABCAAB18599-500
Anti-Protein G Chicken Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Type: Primary
Antigen: Protein G
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR