Tisk…

Anti-CCNA2 Mouse monoclonal antibody unconjugated [clone: E70.1]

Dodavatel: Abcam
ABCAAB18719-50EA 21140 CZK
ABCAAB18719-50
Anti-CCNA2 Mouse monoclonal antibody unconjugated [clone: E70.1]
Protilátky
Type: Primary
Antigen: CCNA2 (cyclin A2)
Clonality: monoclonal
Clone: E70.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR