Tisk…

Anti-RAC1 + Cdc42 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB18758-100EA 21140 CZK
ABCAAB18758-100
Anti-RAC1 + Cdc42 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: RAC1 + Cdc42
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR