Tisk…

Anti-pan-AKT Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB18785-100EA 22860 CZK
ABCAAB18785-100
Anti-pan-AKT Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: pan-AKT
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR