Tisk…

Anti-CTSF Rabbit polyclonal antibody

ABCAAB18858-100EA 16080 CZK
ABCAAB18858-100
Anti-CTSF Rabbit polyclonal antibody
Type: Primary
Antigen: CTSF (cathepsin F)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR