Tisk…

Anti-TGF beta Receptor III Goat Polyclonal Antibody

Dodavatel: Abcam
ABCAAB18885-50EA 22520 CZK
ABCAAB18885-50
Anti-TGF beta Receptor III Goat Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TGF beta Receptor III
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR