Tisk…

Anti-HTRA2 Rabbit polyclonal antibody

ABCAAB18895-100EA 17160 CZK
ABCAAB18895-100
Anti-HTRA2 Rabbit polyclonal antibody
Type: Primary
Antigen: HTRA2 (HtrA serine peptidase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR