Tisk…

Anti-Rsk 2 / MAPKAP Kinase 1b Rabbit polyclonal antibody

ABCAAB18907-100EA 16080 CZK
ABCAAB18907-100
Anti-Rsk 2 / MAPKAP Kinase 1b Rabbit polyclonal antibody
Type: Primary
Antigen: Rsk 2 / MAPKAP Kinase 1b
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR