Tisk…

Anti-CASP3 Rabbit polyclonal antibody unconjugated

ABCAAB49822-100EA 19690 CZK
ABCAAB49822-100
Anti-CASP3 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: CASP3 (caspase 3, apoptosis-related cysteine peptidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR