Tisk…

Anti-CTSK Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB49892-100EA 21540 CZK
ABCAAB49892-100
Anti-CTSK Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CTSK (cathepsin K)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR