Tisk…

Anti-MUC1 Mouse monoclonal antibody unconjugated [clone: E29]

ABCAAB49915-5EA 19690 CZK
ABCAAB49915-5
Anti-MUC1 Mouse monoclonal antibody unconjugated [clone: E29]
Type: Primary
Antigen: MUC1 (mucin 1, cell surface associated)
Clonality: monoclonal
Clone: E29
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR