Tisk…

Anti-PRDX3 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB73349-100EA 31260 CZK
ABCAAB73349-100
Anti-PRDX3 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PRDX3 (peroxiredoxin 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR