Tisk…

Anti-HLCS Rabbit polyclonal antibody

Dodavatel: Abcam
ABCAAB100925-100EA 22800 CZK
ABCAAB100925-100
Anti-HLCS Rabbit polyclonal antibody
Type: Primary
Antigen: HLCS (holocarboxylase synthetase (biotin-(proprionyl-CoA-carboxylase (ATP-hydrolysing)) ligase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR