Tisk…

Anti-STMN1 Rabbit monoclonal antibody [clone: SP49]

Dodavatel: Abcam
ABCAAB101690-100EA 26080 CZK
ABCAAB101690-100
Anti-STMN1 Rabbit monoclonal antibody [clone: SP49]
Type: Primary
Antigen: STMN1 (stathmin 1)
Clonality: monoclonal
Clone: SP49
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR