Tisk…

Anti-S6K Rabbit monoclonal antibody [clone: SP50]

Dodavatel: Abcam
ABCAAB101691-100EA 25920 CZK
ABCAAB101691-100
Anti-S6K Rabbit monoclonal antibody [clone: SP50]
Type: Primary
Antigen: S6K
Clonality: monoclonal
Clone: SP50
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR