Tisk…

Anti-DPP4 Mouse monoclonal antibody [clone: 11D7]

Dodavatel: Abcam
ABCAAB114033-100EA 22030 CZK
ABCAAB114033-100
Anti-DPP4 Mouse monoclonal antibody [clone: 11D7]
Type: Primary
Antigen: DPP4 (dipeptidyl-peptidase 4)
Clonality: monoclonal
Clone: 11D7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR