Tisk…

Anti-Cytokeratin Mouse monoclonal antibody [clone: CK-cocktail]

Dodavatel: Abcam
ABCAAB115959-1000EA 23780 CZK
ABCAAB115959-1000
Anti-Cytokeratin Mouse monoclonal antibody [clone: CK-cocktail]
Type: Primary
Antigen: Cytokeratin
Clonality: monoclonal
Clone: CK-cocktail
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR