Tisk…

Anti-FUT4 Mouse monoclonal antibody [clone: (Workshop IV) (BRA-4F1) our designation 238]

Dodavatel: Abcam
ABCAAB115993-1000EA 23780 CZK
ABCAAB115993-1000
Anti-FUT4 Mouse monoclonal antibody [clone: (Workshop IV) (BRA-4F1) our designation 238]
Type: Primary
Antigen: FUT4 (fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific))
Clonality: monoclonal
Clone: (Workshop IV) (BRA-4F1) our designation 238
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR