Tisk…
Anti-TYRO3 Mouse monoclonal antibody [clone: 5B7]
Dodavatel: Abcam
ABCAAB117877-100EA 24300 CZK
ABCAAB117877-100
Anti-TYRO3 Mouse monoclonal antibody [clone: 5B7]
Type: Primary
Antigen: TYRO3 (TYRO3 protein tyrosine kinase)
Clonality: monoclonal
Clone: 5B7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human