Tisk…

Anti-NMNAT1 Mouse monoclonal antibody [clone: 1F7]

Dodavatel: Abcam
ABCAAB118270-100EA 24300 CZK
ABCAAB118270-100
Anti-NMNAT1 Mouse monoclonal antibody [clone: 1F7]
Type: Primary
Antigen: NMNAT1 (nicotinamide nucleotide adenylyltransferase 1)
Clonality: monoclonal
Clone: 1F7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR