Tisk…

Anti-CYP17A1 Mouse monoclonal antibody [clone: 5G10]

Dodavatel: Abcam
ABCAAB118988-100EA 24850 CZK
ABCAAB118988-100
Anti-CYP17A1 Mouse monoclonal antibody [clone: 5G10]
Type: Primary
Antigen: CYP17A1 (cytochrome P450, family 17, subfamily A, polypeptide 1)
Clonality: monoclonal
Clone: 5G10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR