Tisk…

Anti-PHF21B Mouse monoclonal antibody [clone: 3D1]

Dodavatel: Abcam
ABCAAB119249-100EA 24300 CZK
ABCAAB119249-100
Anti-PHF21B Mouse monoclonal antibody [clone: 3D1]
Type: Primary
Antigen: PHF21B (PHD finger protein 21B)
Clonality: monoclonal
Clone: 3D1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR