Tisk…

Anti-BROX Rabbit polyclonal antibody unconjugated

Dodavatel: Abcam
ABCAAB122174-100EA 25590 CZK
ABCAAB122174-100
Anti-BROX Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: BROX (BRO1 domain and CAAX motif containing)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR