Tisk…
Anti-FCHO2 Rabbit polyclonal antibody unconjugated
Dodavatel: Abcam
ABCAAB122407-100EA 25590 CZK
ABCAAB122407-100
Anti-FCHO2 Rabbit polyclonal antibody unconjugated
Protilátky
Type: Primary
Antigen: FCHO2 (FCH domain only 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human