Tisk…
Anti-ST7 Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB122459-100EA 25010 CZK
ABCAAB122459-100
Anti-ST7 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ST7 (suppression of tumorigenicity 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human