Tisk…
Anti-CDC42SE2 Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB122492-100EA 25010 CZK
ABCAAB122492-100
Anti-CDC42SE2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CDC42SE2 (CDC42 small effector 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human