Tisk…
Anti-C10orf12 Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB122520-100EA 23960 CZK
ABCAAB122520-100
Anti-C10orf12 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: C10orf12 (chromosome 10 open reading frame 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human