Tisk…
Anti-PLAC8 Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB122652-100EA 25560 CZK
ABCAAB122652-100
Anti-PLAC8 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PLAC8 (placenta-specific 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human