Tisk…
Anti-VPS37D Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB122683-100EA 25010 CZK
ABCAAB122683-100
Anti-VPS37D Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: VPS37D (vacuolar protein sorting 37 homolog D (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human