Tisk…

Anti-PTCHD4 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB122705-100EA 25010 CZK
ABCAAB122705-100
Anti-PTCHD4 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PTCHD4 (patched domain containing 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR