Tisk…
Anti-NPVF Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB122738-100EA 25010 CZK
ABCAAB122738-100
Anti-NPVF Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: NPVF (neuropeptide VF precursor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human