Tisk…

Anti-DEFB119 Rabbit polyclonal antibody unconjugated

Dodavatel: Abcam
ABCAAB122826-100EA 25010 CZK
ABCAAB122826-100
Anti-DEFB119 Rabbit polyclonal antibody unconjugated
Protilátky
Type: Primary
Antigen: DEFB119 (defensin, beta 119)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR