Tisk…

Anti-NTN1 Goat Polyclonal Antibody

Dodavatel: Abcam
ABCAAB122903-50EA 25740 CZK
ABCAAB122903-50
Anti-NTN1 Goat Polyclonal Antibody
Protilátky
Type: Primary
Antigen: NTN1 (netrin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR