Tisk…

Anti-NUF2 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB122962-100EA 25680 CZK
ABCAAB122962-100
Anti-NUF2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: NUF2 (NUF2, NDC80 kinetochore complex component)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken, Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR