Tisk…

Anti-MMAB Mouse monoclonal antibody [clone: 2G5]

Dodavatel: Abcam
ABCAAB123916-100EA 23140 CZK
ABCAAB123916-100
Anti-MMAB Mouse monoclonal antibody [clone: 2G5]
Type: Primary
Antigen: MMAB (methylmalonic aciduria (cobalamin deficiency) cblB type)
Clonality: monoclonal
Clone: 2G5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR