Tisk…

Anti-ZFP36 Mouse Monoclonal Antibody [clone: 8B5]

Dodavatel: Abcam
ABCAAB124024-100EA 25560 CZK
ABCAAB124024-100
Anti-ZFP36 Mouse Monoclonal Antibody [clone: 8B5]
Protilátky
Type: Primary
Antigen: ZFP36 (ZFP36 ring finger protein)
Clonality: monoclonal
Clone: 8B5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR