Tisk…
Anti-TNFSF4 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: ATM-2]
Dodavatel: Abcam
ABCAAB124047-50EA 24510 CZK
ABCAAB124047-50
Anti-TNFSF4 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: ATM-2]
Protilátky
Type: Primary
Antigen: TNFSF4 (tumor necrosis factor (ligand) superfamily, member 4)
Clonality: monoclonal
Clone: ATM-2
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat