Tisk…

Anti-Epac1 Rabbit polyclonal antibody unconjugated

Dodavatel: Abcam
ABCAAB124162-100EA 26320 CZK
ABCAAB124162-100
Anti-Epac1 Rabbit polyclonal antibody unconjugated
Protilátky
Type: Primary
Antigen: Epac1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR