Tisk…

Anti-DARC Rabbit polyclonal antibody unconjugated

Dodavatel: Abcam
ABCAAB124648-50EA 25740 CZK
ABCAAB124648-50
Anti-DARC Rabbit polyclonal antibody unconjugated
Protilátky
Type: Primary
Antigen: DARC (Duffy blood group, atypical chemokine receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR