Tisk…

Anti-CBFB Rabbit Monoclonal Antibody [clone: EPR6321]

Dodavatel: Abcam
ABCAAB124693-100EA 25560 CZK
ABCAAB124693-100
Anti-CBFB Rabbit Monoclonal Antibody [clone: EPR6321]
Protilátky
Type: Primary
Antigen: CBFB (core-binding factor, beta subunit)
Clonality: monoclonal
Clone: EPR6321
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR